Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029780.3 | NP_084056.1 | 15, 16 | 2082 | ||
GenBank mRNA | AB052739.1 | - | 2, 3 | 151 | ||
AB057663.2 | - | 15, 16 | 1852 | |||
AK018272.1 | - | 6, 7 | 638 | |||
AK141745.1 | - | 15, 16 | 2083 | |||
AK148327.1 | - | 2479 | ||||
AK191593.1 | - | 1, 2 | 67 | |||
AK193894.1 | - | 1, 2 | 67 | |||
BC015273.1 | - | 15, 16 | 1961 | 4222659 | ||
BC056439.1 | - | 15, 16 | 1999 | |||
BC092040.1 | - | 15, 16 | 1947 | 2645313 |