Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025337.3 | NP_079613.3 | 4 | 689 | ||
GenBank mRNA | AF525358.2 | - | 4 | 591 | ||
AJ271800.1 | - | 4 | 606 | |||
AJ271801.1 | - | 4 | 606 | |||
AK002664.1 | - | 4 | 669 | |||
AK003915.1 | - | 4 | 676 | |||
AK143203.1 | - | 4 | 688 | |||
AK179820.1 | - | 1 | 42 | |||
BC031857.1 | - | 4 | 666 | 5124841 |