Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024206.4 | NP_077168.2 | 7, 8 | 745 | ||
GenBank mRNA | AK003354.1 | - | 7, 8 | 746 | ||
AK009755.1 | - | 6, 7 | 729 | |||
BC002128.1 | - | 7, 8 | 748 | 3590767 |