Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D1Pas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033077.3 | NP_149068.1 | 1 | 1223 | ||
GenBank mRNA | AK029542.1 | - | 1 | 1223 | ||
BC139286.1 | - | 1 | 1218 | 8862308 | ||
BC139287.1 | - | 1 | 1218 | 8862309 | ||
J04847.1 | - | 1 | 763 |