Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053215.3 | NP_444445.2 | 1 | 547 | ||
GenBank mRNA | AK002736.1 | - | 1 | 564 | ||
BC019487.1 | - | 1 | 547 | 4237718 |