Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112703.2 | NP_001106174.1 | 3 | 1628 | ||
NM_001283045.1 | NP_001269974.1 | 3 | 505 | |||
NM_001283046.1 | NP_001269975.1 | 4 | 1833 | |||
NM_001283047.1 | NP_001269976.1 | 4 | 1772 | |||
NM_009594.4 | NP_033724.2 | 3 | 399 | |||
GenBank mRNA | AK088252.1 | - | 4 | 1758 | ||
AK090095.1 | - | 3 | 357 | |||
AK166028.1 | - | 3 | 1617 | |||
AK166210.1 | - | 4 | 1867 | |||
BC059260.1 | - | 3 | 426 | 6408990 | ||
BC103770.1 | - | 3 | 852 | 30848553 | ||
CJ099623.1 | - | 3 | 350 | |||
CJ128107.1 | - | 3 | 394 | |||
CN663033.1 | - | 2 | 120 | |||
J02995.1 | - | 3 | 352 | |||
K03228.1 | - | 3 | 394 | |||
M12263.1 | - | 4 | 759 | |||
M12264.1 | - | 3 | 505 | |||
M12265.1 | - | 3 | 554 |