Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009602.4 | NP_033732.2 | 5 | 609 | ||
GenBank mRNA | AF089739.1 | - | 4 | 299 | ||
AF145286.1 | - | 5 | 548 | |||
AF299083.1 | - | 5 | 403 | |||
AF325349.1 | - | 5 | 369 | |||
AK049722.1 | - | 5 | 582 | |||
AK051742.1 | - | 5 | 591 | |||
AK081254.1 | - | 586 | ||||
AY574267.1 | - | 5 | 383 | |||
BC065103.1 | - | 5 | 562 | 6838257 |