Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009602.4 | NP_033732.2 | 3 | 456 | ||
GenBank mRNA | AF089739.1 | - | 2 | 146 | ||
AF145286.1 | - | 3 | 395 | |||
AF299083.1 | - | 3 | 250 | |||
AF325349.1 | - | 3 | 216 | |||
AK049722.1 | - | 3 | 429 | |||
AK051742.1 | - | 3 | 438 | |||
AK081254.1 | - | 433 | ||||
AY574267.1 | - | 3 | 230 | |||
BC065103.1 | - | 3 | 409 | 6838257 |