Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053248.2 | NP_444478.2 | 9, 10 | 1271 | ||
GenBank mRNA | AF235001.1 | - | 9, 10 | 1238 | ||
AF380353.1 | - | 9, 10 | 1193 | |||
AK046718.1 | - | 9, 10 | 1272 | |||
BC137650.1 | - | 9, 10 | 1206 | 8860669 | ||
BC137651.1 | - | 9, 10 | 1206 | 8860670 |