Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277255.1 | NP_001264184.1 | 10 | 1448 | ||
NM_001277257.1 | NP_001264186.1 | 9 | 1387 | |||
NM_001277258.1 | NP_001264187.1 | 9 | 1356 | |||
NM_001277259.1 | NP_001264188.1 | 10 | 1443 | |||
NM_009612.3 | NP_033742.2 | 9 | 1425 | |||
GenBank mRNA | AK134712.1 | - | 7 | 1182 | ||
AK160915.1 | - | 10 | 1416 | |||
AK170870.1 | - | 7 | 1181 | |||
BC014291.1 | - | 8 | 1344 | 3982004 | ||
BC015083.1 | - | 8 | 1306 | 3980151 | ||
CT010330.1 | - | 7 | 1153 | |||
L48015.1 | - | 9 | 1427 | |||
Z31664.1 | - | 9 | 1393 |