Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271857.1 | NP_001258786.1 | 3 | 530 | ||
NM_001271858.1 | NP_001258787.1 | 3 | 527 | |||
NM_001271859.1 | NP_001258788.1 | 3 | 328 | |||
NM_001271860.1 | NP_001258789.1 | 3 | 530 | |||
NM_001271861.1 | NP_001258790.1 | 3 | 331 | |||
NM_013458.5 | NP_038486.2 | 3 | 331 | |||
GenBank mRNA | AF100422.1 | - | 2 | 206 | ||
AF100423.1 | - | 2 | 206 | |||
AF189769.1 | - | 1 | 128 | |||
AF189770.1 | - | 1 | 128 | |||
AK014496.1 | - | 3 | 296 | |||
AK028425.1 | - | 3 | 302 | |||
AK029488.1 | - | 3 | 481 | |||
AK147544.1 | - | 3 | 403 | |||
AK156954.1 | - | 3 | 320 | |||
AK160736.1 | - | 3 | 484 | |||
BC046783.1 | - | 3 | 464 | 6401200 | ||
BC053032.1 | - | 3 | 464 | 6401200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK138207.1 | - | 728 | Chr. - - [-] on Build |