Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adra1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271759.1 | NP_001258688.1 | 2, 3 | 1837 | ||
NM_001271760.1 | NP_001258689.1 | 2, 3 | 1364 | |||
NM_013461.4 | NP_038489.2 | 2, 3 | 1827 | |||
GenBank mRNA | AF031431.1 | - | 1, 2 | 887 | ||
AK079597.1 | - | 2, 3 | 1820 | |||
AK085653.1 | - | 2, 3 | 1839 | |||
AK138919.1 | - | 2, 3 | 1748 | |||
BC113139.1 | - | 2, 3 | 1059 | 40048262 |
Set Membership: |
G-protein coupled receptor |