Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035145.1 | - | 13 | 1652 | ||
AK042326.1 | - | 13 | 1656 | |||
AK078626.1 | - | 13 | 1653 | |||
AK079602.1 | - | 13 | 1627 | |||
AK128913.1 | - | 13 | 1589 | |||
AK146286.1 | - | 13 | 1645 | |||
AK150233.1 | - | 13 | 1644 | |||
AK150250.1 | - | 13 | 1655 | |||
AK150920.1 | - | 13 | 1644 | |||
AK150986.1 | - | 13 | 1626 | |||
AK151763.1 | - | 13 | 1644 | |||
AK151826.1 | - | 13 | 1629 | |||
AK153128.1 | - | 13 | 1626 | |||
AK160060.1 | - | 13 | 1646 | |||
AK163452.1 | - | 1655 | ||||
AK165032.1 | - | 13 | 1890 | |||
AK184586.1 | - | 3 | 202 | |||
AK188389.1 | - | 3 | 235 | |||
AK189526.1 | - | 3 | 203 | |||
AK206041.1 | - | 4 | 376 | |||
BC005476.1 | - | 13 | 1634 | 3600875 | ||
S71509.1 | - | 13 | 1605 | |||
U07235.1 | - | 13 | 1651 |