Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023040.3 | NP_075527.2 | 3 | 593 | ||
GenBank mRNA | AB041561.1 | - | 3 | 510 | ||
AF148688.1 | - | 3 | 373 | |||
AK146579.1 | - | 3 | 594 | |||
BC023941.1 | - | 3 | 593 | 5342526 | ||
BI689446.1 | - | 1 | 72 | |||
CA321838.1 | - | 1 | 58 |