Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012363.2 | NP_001012363.2 | 4 | 422 | ||
NM_001102414.1 | NP_001095884.1 | 4 | 621 | |||
NM_001102415.1 | NP_001095885.1 | 4 | 621 | |||
NM_145559.2 | NP_663534.1 | 5 | 616 | |||
GenBank mRNA | AF469480.1 | - | 4 | 625 | ||
AF490463.1 | - | 4 | 446 | |||
AK037093.1 | - | 4 | 474 | |||
AK156740.1 | - | 4 | 525 | |||
AK172234.1 | - | 4 | 525 | |||
AY776155.1 | - | 4 | 401 | |||
BC006076.1 | - | 5 | 616 | 3593581 | ||
BC138213.1 | - | 5 | 557 | 8861232 | ||
BC145198.1 | - | 5 | 557 | 9053749 |