Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009704.3 | NP_033834.1 | 1, 2 | 254 | ||
GenBank mRNA | AK018590.1 | - | 1, 2 | 270 | ||
AK020273.1 | - | 1, 2 | 270 | |||
BC009138.1 | - | 1, 2 | 254 | 3597695 | ||
CT010320.1 | - | 1, 2 | 55 | |||
D12648.1 | - | 1, 2 | 81 |