Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031259.1 | - | 2 | 201 | ||
AK042977.1 | - | 2 | 241 | |||
AK053123.1 | - | 2 | 659 | |||
AK077591.1 | - | 2 | 214 | |||
AK167133.1 | - | 2 | 211 | |||
AK167601.1 | - | 2 | 215 | |||
AK167653.1 | - | 2 | 214 | |||
AK169094.1 | - | 2 | 211 | |||
AK169412.1 | - | 2 | 211 | |||
BC010487.1 | - | 2 | 494 | 3483748 | ||
BY129623.1 | - | 2 | 253 | |||
D87899.1 | - | 2 | 206 |