Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301301.1 | NP_001288230.1 | 6 | 669 | ||
NM_001301303.1 | NP_001288232.1 | 6 | 656 | |||
NM_001301305.1 | NP_001288234.1 | 6 | 648 | |||
NM_001301308.1 | NP_001288237.1 | 5 | 498 | |||
NM_001301309.1 | NP_001288238.1 | 5 | 479 | |||
NM_007486.5 | NP_031512.1 | 6 | 688 | |||
GenBank mRNA | AK002516.1 | - | 6 | 650 | ||
AK008273.1 | - | 6 | 615 | |||
AK169733.1 | - | 6 | 646 | |||
AW543335.1 | - | 1 | 451 | |||
BC031763.1 | - | 6 | 601 | 4923219 | ||
CO038885.1 | - | 1 | 512 | |||
L07918.1 | - | 6 | 602 |