Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301301.1 | NP_001288230.1 | 5, 6 | 496 | ||
NM_001301303.1 | NP_001288232.1 | 5, 6 | 483 | |||
NM_001301305.1 | NP_001288234.1 | 5, 6 | 475 | |||
NM_001301308.1 | NP_001288237.1 | 4, 5 | 325 | |||
NM_001301309.1 | NP_001288238.1 | 4, 5 | 306 | |||
NM_007486.5 | NP_031512.1 | 5, 6 | 515 | |||
GenBank mRNA | AA219822.1 | - | 5, 6 | 449 | ||
AK002516.1 | - | 5, 6 | 477 | |||
AK008273.1 | - | 5, 6 | 442 | |||
AK169733.1 | - | 5, 6 | 473 | |||
BB755757.1 | - | 5, 6 | 465 | |||
BC031763.1 | - | 5, 6 | 428 | 4923219 | ||
L07918.1 | - | 5, 6 | 429 |