Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019734.2 | NP_062708.1 | 5 | 471 | ||
GenBank mRNA | AF157500.1 | - | 5 | 401 | ||
AF352179.1 | - | 5 | 413 | |||
AK075666.1 | - | 5 | 386 | |||
AK075919.1 | - | 5 | 425 | |||
AK077721.1 | - | 5 | 424 | |||
AK090314.1 | - | 5 | 421 | |||
AK150567.1 | - | 5 | 384 | |||
AK151208.1 | - | 5 | 472 | |||
AK151312.1 | - | 5 | 384 | |||
AK151478.1 | - | 5 | 462 | |||
AK152305.1 | - | 5 | 423 | |||
AK152811.1 | - | 5 | 384 | |||
AK152920.1 | - | 5 | 431 | |||
AK153101.1 | - | 5 | 431 | |||
AK153297.1 | - | 5 | 385 | |||
AK159556.1 | - | 5 | 393 | |||
AK166757.1 | - | 5 | 403 | |||
BC003204.1 | - | 5 | 391 | 3587042 |