Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019734.2 | NP_062708.1 | 11 | 938 | ||
GenBank mRNA | AF157500.1 | - | 11 | 868 | ||
AF352179.1 | - | 11 | 880 | |||
AK075666.1 | - | 11 | 853 | |||
AK075919.1 | - | 11 | 892 | |||
AK077721.1 | - | 11 | 891 | |||
AK090314.1 | - | 11 | 888 | |||
AK150567.1 | - | 11 | 850 | |||
AK151208.1 | - | 11 | 939 | |||
AK151312.1 | - | 11 | 850 | |||
AK151478.1 | - | 11 | 929 | |||
AK152305.1 | - | 11 | 890 | |||
AK152811.1 | - | 11 | 851 | |||
AK152920.1 | - | 11 | 898 | |||
AK153101.1 | - | 11 | 898 | |||
AK153297.1 | - | 11 | 852 | |||
AK159556.1 | - | 11 | 860 | |||
AK166757.1 | - | 11 | 870 | |||
BC003204.1 | - | 11 | 858 | 3587042 |