Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurod6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009717.2 | NP_033847.1 | 2 | 1266 | ||
GenBank mRNA | AK144303.1 | - | 2 | 1048 | ||
AK144360.1 | - | 2 | 1267 | |||
AK144377.1 | - | 2 | 1100 | |||
BC087831.1 | - | 2 | 1059 | 6411826 | ||
U29086.1 | - | 2 | 1072 |