Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144900.2 | NP_659149.1 | 13, 14 | 2128 | ||
GenBank mRNA | AK088000.1 | - | 13, 14 | 2089 | ||
AK137001.1 | - | 13, 14 | 2087 | |||
AK150523.1 | - | 6, 7 | 1033 | |||
AK159279.1 | - | 13, 14 | 2085 | |||
AK159306.1 | - | 13, 14 | 2083 | |||
AK184412.1 | - | 1, 2 | 185 | |||
AK199691.1 | - | 1, 2 | 187 | |||
AK211215.1 | - | 1, 2 | 185 | |||
BC010319.1 | - | 11, 12 | 1713 | |||
BC021496.1 | - | 13, 14 | 2078 | 5346159 | ||
BC023794.1 | - | 14, 15 | 2023 | 5351943 | ||
BC023964.1 | - | 13, 14 | 2069 | |||
BC025037.1 | - | 6, 7 | 928 | |||
BC025618.1 | - | 13, 14 | 2087 | 5320976 | ||
BC025627.1 | - | 13, 14 | 2078 | 5320513 | ||
BC025811.1 | - | 13, 14 | 2065 | 5096872 | ||
BC030326.1 | - | 2, 3 | 425 | |||
BC032187.1 | - | 13, 14 | 2128 | 5321268 | ||
BC033435.1 | - | 13, 14 | 2024 | 4950746 | ||
BC033471.1 | - | 13, 14 | 2068 | 4954119 | ||
BC034177.1 | - | 12, 13 | 1833 | |||
BC035352.1 | - | 12, 13 | 1798 | |||
BC042435.1 | - | 13, 14 | 2078 | 4951288 |