Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007531.2 | NP_031557.2 | 7, 8 | 946 | ||
GenBank mRNA | AK002334.1 | - | 7, 8 | 912 | ||
AK011107.1 | - | 7, 8 | 913 | |||
AK090088.1 | - | 7, 8 | 903 | |||
AK132052.1 | - | 7, 8 | 908 | |||
AK132058.1 | - | 7, 8 | 935 | |||
AK135729.1 | - | 7, 8 | 863 | |||
AK151952.1 | - | 7, 8 | 940 | |||
AK152375.1 | - | 6, 7 | 1595 | |||
AK153368.1 | - | 6, 7 | 1595 | |||
AY211613.1 | - | 7, 8 | 908 | |||
AY319418.1 | - | 7, 8 | 798 | |||
BC145875.1 | - | 7, 8 | 898 | 40131178 | ||
X78683.1 | - | 7, 8 | 902 |