Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009745.2 | NP_033875.2 | 1, 2 | 216 | ||
GenBank mRNA | AJ011145.1 | - | 1, 2 | 96 | ||
AK131643.1 | - | 1, 2 | 112 | |||
AK160439.1 | - | 1, 2 | 199 | |||
AK186334.1 | - | 1, 2 | 45 | |||
BC011471.1 | - | 1, 2 | 193 | 3660754 | ||
BY087167.1 | - | 1, 2 | 216 |