Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009748.2 | NP_033878.1 | 3 | 291 | ||
GenBank mRNA | AF007552.1 | - | 3 | 279 | ||
AK078796.1 | - | 3 | 287 | |||
BC005572.1 | - | 3 | 291 | 3967968 |