Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 8, 9 | 1026 | ||
GenBank mRNA | AK034049.1 | - | 8, 9 | 1005 | ||
AK034966.1 | - | 827 | ||||
AK042069.1 | - | 8, 9 | 996 | |||
AK048460.1 | - | 8, 9 | 957 | |||
AK076517.1 | - | 8, 9 | 997 | |||
AK153218.1 | - | 8, 9 | 1008 | |||
AK166473.1 | - | 8, 9 | 1011 | |||
AK167271.1 | - | 6, 7 | 636 | |||
AK171593.1 | - | 8, 9 | 1029 | |||
AK171760.1 | - | 7, 8 | 827 | |||
BC003998.1 | - | 6, 7 | 597 | |||
BC028875.1 | - | 8, 9 | 946 | 4240285 | ||
M57734.1 | - | 8, 9 | 985 |