Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 11, 12 | 1264 | ||
GenBank mRNA | AK034966.1 | - | 1156 | |||
AK042069.1 | - | 11, 12 | 1234 | |||
AK048460.1 | - | 11, 12 | 1195 | |||
AK076517.1 | - | 11, 12 | 1235 | |||
AK153218.1 | - | 11, 12 | 1246 | |||
AK166473.1 | - | 11, 12 | 1250 | |||
AK171593.1 | - | 11, 12 | 1267 | |||
AK171760.1 | - | 10, 11 | 1065 | |||
AK204903.1 | - | 3, 4 | 242 | |||
AK217231.1 | - | 2, 3 | 210 | |||
BC003998.1 | - | 9, 10 | 835 | |||
BC028875.1 | - | 11, 12 | 1184 | 4240285 | ||
M57734.1 | - | 11, 12 | 1223 |