Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009754.3 | NP_033884.1 | 4 | 496 | ||
NM_207680.2 | NP_997563.1 | 4 | 754 | |||
NM_207681.2 | NP_997564.1 | 5 | 586 | |||
GenBank mRNA | AF032459.1 | - | 3 | 526 | ||
AF032460.1 | - | 4 | 358 | |||
AF032461.1 | - | 3 | 268 | |||
AK011490.1 | - | 4 | 743 | |||
AK082613.1 | - | 4 | 750 | |||
AK150464.1 | - | 4 | 497 | |||
AK152472.1 | - | 4 | 497 | |||
AK153542.1 | - | 4 | 497 | |||
BC019556.1 | - | 4 | 748 | 4459720 |