Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147778.3 | NP_680087.1 | 3 | 279 | ||
GenBank mRNA | AK007318.1 | - | 3 | 284 | ||
AK041634.1 | - | 3 | 280 | |||
AK145955.1 | - | 3 | 307 | |||
AK145999.1 | - | 3 | 309 | |||
AK163947.1 | - | 2 | 1577 | |||
BC038077.1 | - | 3 | 321 | 4013160 | ||
BC048815.1 | - | 3 | 290 | 3482368 |