Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110504.1 | NP_001103974.1 | 10 | 1137 | ||
NM_007600.3 | NP_031626.1 | 10 | 1189 | |||
GenBank mRNA | AF021847.1 | - | 10 | 1068 | ||
AF084459.1 | - | 9 | 1055 | |||
AK088547.1 | - | 10 | 1109 | |||
AK149090.1 | - | 6 | 537 | |||
AK149706.1 | - | 10 | 1129 | |||
AK150084.1 | - | 10 | 1128 | |||
AK160940.1 | - | 10 | 1129 | |||
AK165258.1 | - | 10 | 1148 | |||
AK168063.1 | - | 10 | 1128 | |||
AK171405.1 | - | 10 | 1092 | |||
BC026138.1 | - | 10 | 1104 | 4218148 | ||
BC050276.1 | - | 10 | 1105 | 4923347 |