Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110504.1 | NP_001103974.1 | 7 | 865 | ||
NM_007600.3 | NP_031626.1 | 7 | 917 | |||
GenBank mRNA | AF021847.1 | - | 7 | 796 | ||
AF084459.1 | - | 6 | 783 | |||
AK088547.1 | - | 7 | 837 | |||
AK149090.1 | - | 3 | 265 | |||
AK149706.1 | - | 7 | 857 | |||
AK150084.1 | - | 7 | 856 | |||
AK160940.1 | - | 7 | 857 | |||
AK165258.1 | - | 7 | 876 | |||
AK168063.1 | - | 7 | 856 | |||
AK171405.1 | - | 7 | 820 | |||
AK196291.1 | - | 3 | 301 | |||
BC026138.1 | - | 7 | 832 | 4218148 | ||
BC050276.1 | - | 7 | 833 | 4923347 |