Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037761.2 | NP_001032850.1 | 5 | 497 | ||
NM_001271405.1 | NP_001258334.1 | 5 | 476 | |||
NM_001271406.1 | NP_001258335.1 | 5 | 462 | |||
NM_009798.4 | NP_033928.1 | 5 | 497 | |||
GenBank mRNA | AK156778.1 | - | 5 | 466 | ||
AK159390.1 | - | 5 | 471 | |||
AK160082.1 | - | 5 | 472 | |||
AK162757.1 | - | 5 | 464 | |||
AK168738.1 | - | 5 | 425 | |||
BC002053.1 | - | 5 | 439 | 3590980 | ||
FJ692320.1 | - | 5 | 462 | |||
U10406.1 | - | 5 | 417 | |||
U10407.1 | - | 5 | 388 |