Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011710.1 | - | 4 | 291 | ||
AK146363.1 | - | 4 | 301 | |||
AK164321.1 | - | 4 | 267 | |||
AK165998.1 | - | 4 | 278 | |||
AK168842.1 | - | 4 | 299 | |||
BC002022.1 | - | 4 | 287 | 3491126 | ||
BY070840.1 | - | 4 | 327 | |||
BY265671.1 | - | 4 | 337 | |||
Y13087.1 | - | 3 | 255 |