Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007622.3 | NP_031648.1 | 4 | 653 | ||
GenBank mRNA | AK014215.1 | - | 4 | 629 | ||
AK017608.1 | - | 4 | 542 | |||
AK157710.1 | - | 4 | 655 | |||
BC055290.1 | - | 4 | 596 | 6313666 | ||
X56690.1 | - | 3 | 398 |