Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007630.2 | NP_031656.2 | 2 | 252 | ||
GenBank mRNA | AK013312.1 | - | 2 | 221 | ||
AK076122.1 | - | 2 | 246 | |||
BC008247.1 | - | 2 | 168 | 3156950 | ||
BY300844.1 | - | 2 | 255 | |||
CT010286.1 | - | 2 | 138 | |||
X66032.1 | - | 2 | 239 |