Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111099.1 | NP_001104569.1 | 2 | 175 | ||
NM_007669.4 | NP_031695.1 | 2 | 149 | |||
GenBank mRNA | AB017817.1 | - | 1 | 48 | ||
AB017818.1 | - | 1 | 48 | |||
AK007630.1 | - | 2 | 147 | |||
AK089093.1 | - | 2 | 147 | |||
AK139306.1 | - | 2 | 150 | |||
AK149910.1 | - | 2 | 150 | |||
AK170467.1 | - | 2 | 150 | |||
BC002043.1 | - | 2 | 102 | 3495942 | ||
BY206886.1 | - | 2 | 175 | |||
CT010218.1 | - | 1 | 48 | |||
U09507.1 | - | 1 | 248 | |||
U24173.1 | - | 2 | 140 |
Gene Symbol: 4930567H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131763.1 | - | 431 |