Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007672.2 | NP_031698.2 | 5 | 1180 | ||
GenBank mRNA | AK148057.1 | - | 5 | 1164 | ||
AK171706.1 | - | 5 | 1135 | |||
AK187086.1 | - | 1 | 153 | |||
BC117530.1 | - | 5 | 1069 | 40106297 | ||
BC118511.1 | - | 5 | 1069 | 40106298 | ||
U88588.1 | - | 6 | 1073 |