Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009931.2 | NP_034061.2 | 44 | 4133 | ||
GenBank mRNA | AK146487.1 | - | 44 | 4082 | ||
AK147284.1 | - | 44 | 4055 | |||
AK147355.1 | - | 44 | 4058 | |||
AK147661.1 | - | 44 | 4099 | |||
AK159130.1 | - | 44 | 4095 | |||
BC056620.1 | - | 3 | 368 | |||
BC072650.1 | - | 44 | 3996 | 6852661 | ||
J04694.1 | - | 44 | 4037 | |||
M14042.1 | - | 5 | 462 | |||
X02201.1 | - | 2 | 81 |