Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293559.1 | NP_001280488.1 | 3, 4 | 293 | ||
NM_009941.3 | NP_034071.2 | 3, 4 | 368 | |||
GenBank mRNA | AK002943.1 | - | 625 | |||
AK012583.1 | - | 3, 4 | 305 | |||
AK019276.1 | - | 3, 4 | 270 | |||
AK150447.1 | - | 3, 4 | 298 | |||
AV469576.1 | - | 3, 4 | 293 | |||
BC132269.1 | - | 3, 4 | 289 | 40130546 | ||
BC132275.1 | - | 3, 4 | 289 | 40130552 | ||
BY250039.1 | - | 3, 4 | 371 | |||
M37829.1 | - | 3, 4 | 291 | |||
X54691.1 | - | 3, 4 | 293 |