Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009999.4 | NP_034129.1 | 4, 5 | 666 | ||
GenBank mRNA | AF128849.1 | - | 4, 5 | 648 | ||
AF129405.1 | - | 3, 4 | 462 | |||
AK028103.1 | - | 4, 5 | 666 | |||
BC060973.1 | - | 4, 5 | 660 | 30301385 | ||
M21856.1 | - | 4, 5 | 648 | |||
X99715.1 | - | 4, 5 | 644 |