Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010005.3 | NP_034135.2 | 9 | 1509 | ||
GenBank mRNA | AA239803.1 | - | 2 | 206 | ||
AI323751.1 | - | 1 | 134 | |||
BC010989.1 | - | 9 | 1507 | 4193743 | ||
BC051039.1 | - | 8 | 1344 | |||
BC057924.1 | - | 9 | 1496 | 5100459 | ||
M27167.1 | - | 9 | 1487 |