Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013741.1 | NP_001013763.1 | 2 | 1571 | ||
GenBank mRNA | AK122363.1 | - | 2 | 1446 | ||
AK158894.1 | - | 2 | 1572 | |||
AK164154.1 | - | 1 | 1180 | |||
BC157995.1 | - | 2 | 1317 | 9088145 | ||
BC157996.1 | - | 2 | 1317 | 9088146 |