Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110826.1 | NP_001104296.1 | 13 | 1765 | ||
NM_007841.4 | NP_031867.1 | 13 | 1708 | |||
NM_181324.3 | NP_851841.2 | 13 | 1733 | |||
GenBank mRNA | AF038995.1 | - | 12 | 1327 | ||
AK054144.1 | - | 13 | 1727 | |||
AK148483.1 | - | 13 | 1730 | |||
BC021452.1 | - | 12 | 1625 | 5043599 |