Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190490.1 | NP_001177419.1 | 5 | 674 | ||
NM_001190491.1 | NP_001177420.1 | 5 | 674 | |||
NM_032418.2 | NP_115794.1 | 5 | 674 | |||
GenBank mRNA | BC024150.1 | - | 5 | 661 | 4217643 | |
BC056615.1 | - | 5 | 653 | |||
BC075715.1 | - | 5 | 582 | 4989259 | ||
S60313.1 | - | 2 | 181 | |||
S60314.1 | - | 2 | 181 | |||
S60315.1 | - | 2 | 181 | |||
S60316.1 | - | 2 | 181 |