Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290709.1 | NP_001277638.1 | 11, 12 | 1201 | ||
NM_001290710.1 | NP_001277639.1 | 11, 12 | 1174 | |||
NM_001290711.1 | NP_001277640.1 | 11, 12 | 1317 | |||
NM_007897.3 | NP_031923.1 | 11, 12 | 1198 | |||
GenBank mRNA | AK031239.1 | - | 11, 12 | 1196 | ||
AK035781.1 | - | 11, 12 | 1081 | |||
AK137389.1 | - | 11, 12 | 1239 | |||
AK139021.1 | - | 11, 12 | 1172 | |||
AK141175.1 | - | 11, 12 | 1201 | |||
BC139362.1 | - | 11, 12 | 1159 | 8862384 | ||
BC139364.1 | - | 11, 12 | 1159 | 8862386 | ||
BC139765.1 | - | 4, 5 | 650 | |||
L12147.1 | - | 11, 12 | 1164 |