Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012266.1 | - | 2 | 402 | ||
AK027977.1 | - | 2 | 374 | |||
BC004620.1 | - | 2 | 380 | 3585323 | ||
BC004703.1 | - | 2 | 1012 | 3499265 | ||
X97755.1 | - | 2 | 378 |