Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010119.5 | NP_034249.1 | 5 | 1534 | ||
GenBank mRNA | AF099186.1 | - | 5 | 1537 | ||
AF173156.1 | - | 5 | 1545 | |||
AK012896.1 | - | 5 | 1510 | |||
AK077536.1 | - | 5 | 1514 | |||
AK088980.1 | - | 6 | 1676 | |||
AK152821.1 | - | 5 | 1482 | |||
AK154651.1 | - | 5 | 1523 | |||
AK159191.1 | - | 5 | 1509 | |||
AK159943.1 | - | 5 | 1519 | |||
AK160535.1 | - | 5 | 1522 | |||
AK166701.1 | - | 5 | 1520 | |||
AK167721.1 | - | 5 | 1526 | |||
AK168610.1 | - | 5 | 1510 | |||
AK170679.1 | - | 5 | 1310 | |||
AK171047.1 | - | 5 | 1538 | |||
AK172539.1 | - | 5 | 1510 | |||
AK186064.1 | - | 1 | 287 | |||
AK213323.1 | - | 1 | 297 | |||
BC018268.1 | - | 1 | 295 | |||
BC037094.1 | - | 5 | 1521 | |||
BC043332.1 | - | 5 | 1504 | 4981832 | ||
BC054828.1 | - | 5 | 1498 | 5042951 |