Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133660.3 | NP_598421.1 | 5 | 814 | ||
GenBank mRNA | AK144549.1 | - | 5 | 799 | ||
BC019208.1 | - | 5 | 745 | 5052893 | ||
S80191.1 | - | 5 | 744 |