Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023670.3 | NP_076159.3 | 12 | 1832 | ||
GenBank mRNA | AB046173.1 | - | 12 | 1349 | ||
AK011689.1 | - | 12 | 1613 | |||
AK045244.1 | - | 2 | 542 | |||
AK088465.1 | - | 12 | 1591 | |||
AK147140.1 | - | 12 | 1836 | |||
BC045138.1 | - | 12 | 1462 | 6306448 | ||
BC049082.1 | - | 12 | 1498 | 5706238 |